Assay Details
| Gene Symbol: BUB3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756675.3 | XP_002756721.1 | 6 - 7 | 988 | 111 | |
| XM_003734551.3 | XP_003734599.1 | 6 - 7 | 988 | 111 | ||
| GenBank mRNA | EF215267.1 | - | 1 - 2 | 186 | 111 | |
More Information
Set Membership: |
|

BUB3
Genomic Map