Assay Details
| Gene Symbol: SNRK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758253.2 | XP_002758299.1 | 5 - 6 | 1116 | 67 | |
| XM_002758254.1 | XP_002758300.1 | 4 - 5 | 1066 | 67 | ||
| XM_002758255.1 | XP_002758301.1 | 3 - 4 | 1004 | 67 | ||
| XM_008981480.2 | XP_008979728.1 | 4 - 5 | 1014 | 67 | ||
| XM_017964823.1 | XP_017820312.1 | 3 - 4 | 3071 | 67 | ||
More Information
Set Membership: |
|

SNRK
Genomic Map