Assay Details
| Gene Symbol: AKR1C1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008999856.1 | XP_008998104.1 | 12 - 13 | 1812 | 131 | |
| XM_017974016.1 | XP_017829505.1 | 11 - 12 | 1605 | 131 | ||
| XM_017974017.1 | XP_017829506.1 | 10 - 11 | 1367 | 131 | ||
| XM_017974018.1 | XP_017829507.1 | 10 - 11 | 1316 | 131 | ||
More Information
Set Membership: |
|

AKR1C1
Genomic Map