Assay Details
| Gene Symbol: GALNT17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743912.3 | XP_002743958.1 | 4 - 5 | 1197 | 65 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

GALNT17| Gene Symbol: GALNT17 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743912.3 | XP_002743958.1 | 4 - 5 | 1197 | 65 | |
Set Membership: |
|