Assay Details
| Gene Symbol: PUS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753163.3 | XP_002753209.1 | 4 - 5 | 913 | 80 | |
| XM_009004918.2 | XP_009003166.1 | 4 - 5 | 802 | 80 | ||
| XM_009004920.2 | XP_009003168.1 | 4 - 5 | 528 | 80 | ||
| XM_009004921.2 | XP_009003169.1 | 3 - 4 | 712 | 80 | ||
More Information
Set Membership: |
|

PUS1
Genomic Map