Assay Details
Gene Symbol: ZNF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_008982797.2 | XP_008981045.1 | 3 - 4 | 365 | 86 | |
XM_017966462.1 | XP_017821951.1 | 3 - 4 | 437 | 86 | ||
XM_017966463.1 | XP_017821952.1 | 3 - 4 | 659 | 86 | ||
XM_017966467.1 | XP_017821956.1 | 1 - 2 | 318 | 86 |
