Assay Details
| Gene Symbol: C11H11orf58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755001.4 | XP_002755047.1 | 4 - 5 | 638 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

C11H11orf58| Gene Symbol: C11H11orf58 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755001.4 | XP_002755047.1 | 4 - 5 | 638 | 64 | |
Set Membership: |
|