Assay Details
Gene Symbol: SPACA9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002743421.3 | XP_002743467.2 | 3 - 4 | 551 | 72 | |
XM_009004563.2 | XP_009002811.1 | 4 - 5 | 521 | 72 | ||
XM_009004565.2 | XP_009002813.1 | 4 - 5 | 1006 | 72 | ||
XM_009004570.2 | XP_009002818.1 | 4 - 5 | 1010 | 72 | ||
XM_009004593.2 | XP_009002841.1 | 5 - 6 | 707 | 72 | ||
XM_009004603.2 | XP_009002851.1 | 3 - 4 | 954 | 72 | ||
XM_009004610.2 | XP_009002858.1 | 4 - 5 | 531 | 72 | ||
XM_009004617.2 | XP_009002865.1 | 4 - 5 | 603 | 72 | ||
XM_009004622.2 | XP_009002870.1 | 4 - 5 | 607 | 72 | ||
XM_017977047.1 | XP_017832536.1 | 5 - 6 | 1194 | 72 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
