Assay Details
| Gene Symbol: FKBP1B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757993.3 | XP_002758039.1 | 2 - 3 | 242 | 87 | |
| XM_008981074.1 | XP_008979322.1 | 2 - 3 | 251 | 87 | ||
| XM_008981075.2 | XP_008979323.1 | 2 - 3 | 142 | 87 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008981075.2 transcripts.
Set Membership: |
|

FKBP1B
Genomic Map