Assay Details
| Gene Symbol: LOC100402504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753648.3 | XP_002753694.1 | 3 - 4 | 616 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100402504| Gene Symbol: LOC100402504 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002753648.3 | XP_002753694.1 | 3 - 4 | 616 | 86 | |
Set Membership: |
|