Assay Details
| Gene Symbol: ZNF621 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008984069.2 | XP_008982317.1 | 4 - 5 | 2002 | 65 | |
| XM_008984070.2 | XP_008982318.1 | 4 - 5 | 703 | 65 | ||
| XM_008984071.2 | XP_008982319.1 | 4 - 5 | 362 | 65 | ||
| XM_008984073.2 | XP_008982321.1 | 4 - 5 | 1999 | 65 | ||
| XM_008984075.2 | XP_008982323.1 | 4 - 5 | 420 | 65 | ||
| XM_017965972.1 | XP_017821461.1 | 5 - 6 | 1956 | 65 | ||
| XM_017965973.1 | XP_017821462.1 | 5 - 6 | 1374 | 65 | ||
| XM_017965974.1 | XP_017821463.1 | 4 - 5 | 1999 | 65 | ||
More Information
Set Membership: |
|

ZNF621
Genomic Map