Assay Details
| Gene Symbol: TMED3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749215.3 | XP_002749261.1 | 1 - 2 | 360 | 72 | |
| XM_017973337.1 | XP_017828826.1 | 1 - 2 | 353 | 72 | ||
More Information
Set Membership: |
|

TMED3
Genomic Map