Assay Details
| Gene Symbol: MBOAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017970865.1 | XP_017826354.1 | 7 - 8 | 1124 | 67 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MBOAT1| Gene Symbol: MBOAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017970865.1 | XP_017826354.1 | 7 - 8 | 1124 | 67 | |
Set Membership: |
|