Assay Details
Gene Symbol: DPM1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002747659.4 | XP_002747705.2 | 2 - 3 | 300 | 71 | |
XM_008996130.2 | XP_008994378.1 | 2 - 3 | 301 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
