Assay Details
| Gene Symbol: NTF4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008988362.2 | XP_008986610.1 | 2 - 3 | 247 | 65 | |
| XM_017967400.1 | XP_017822889.1 | 2 - 3 | 502 | 65 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_008988362.2, XM_017967400.1 transcripts.
Set Membership: |
|

NTF4
Genomic Map