Assay Details
| Gene Symbol: CHEK2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009008587.2 | XP_009006835.1 | 15 - 16 | 1906 | 71 | |
| XM_017978763.1 | XP_017834252.1 | 16 - 17 | 1976 | 71 | ||
| XM_017978771.1 | XP_017834260.1 | 15 - 16 | 1834 | 71 | ||
| XM_017978772.1 | XP_017834261.1 | 16 - 17 | 2002 | 71 | ||
| XM_017978778.1 | XP_017834267.1 | 15 - 16 | 1876 | 71 | ||
| XM_017978780.1 | XP_017834269.1 | 15 - 16 | 1873 | 71 | ||
| XM_017978785.1 | XP_017834274.1 | 14 - 15 | 1819 | 71 | ||
| XM_017978787.1 | XP_017834276.1 | 14 - 15 | 1789 | 71 | ||
More Information
Set Membership: |
|

CHEK2
Genomic Map