Assay Details
| Gene Symbol: TYMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002806574.3 | XP_002806620.1 | 4 - 5 | 523 | 84 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TYMP| Gene Symbol: TYMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002806574.3 | XP_002806620.1 | 4 - 5 | 523 | 84 | |
Set Membership: |
|