Assay Details
| Gene Symbol: APOC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002754445.4 | XP_002754491.1 | 2 - 3 | 125 | 93 | |
| XM_009006851.2 | XP_009005099.1 | 2 - 3 | 234 | 93 | ||
More Information
Set Membership: |
|

APOC3
Genomic Map