Assay Details
| Gene Symbol: RHCE | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_003733373.3 | XP_003733421.1 | 4 - 5 | 773 | 64 | |
| XM_009000656.2 | XP_008998904.1 | 5 - 6 | 730 | 64 | ||
| XM_009000657.2 | XP_008998905.1 | 4 - 5 | 716 | 64 | ||
| XM_009000658.2 | XP_008998906.1 | 3 - 4 | 692 | 64 | ||
| XM_009000659.2 | XP_008998907.1 | 4 - 5 | 773 | 64 | ||
| XM_009000660.2 | XP_008998908.1 | 4 - 5 | 773 | 64 | ||
| XM_017974419.1 | XP_017829908.1 | 4 - 5 | 691 | 64 | ||
| XM_017974420.1 | XP_017829909.1 | 4 - 5 | 676 | 64 | ||
| GenBank mRNA | AF012429.1 | - | 4 - 5 | 658 | 64 | |
More Information
Set Membership: |
|

RHCE
Genomic Map