Assay Details
| Gene Symbol: IL4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001257228.1 | NP_001244157.1 | 3 - 4 | 430 | 66 | |
| GenBank mRNA | EF493341.1 | - | 3 - 4 | 430 | 66 | |
More Information
Set Membership: |
|

IL4
Genomic Map