Assay Details
| Gene Symbol: PRKACA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987412.2 | XP_008985660.1 | 536 | 66 | ||
| XM_017967752.1 | XP_017823241.1 | 347 | 66 | |||
| XM_017967753.1 | XP_017823242.1 | 1145 | 66 | |||
More Information
Set Membership: |
|

PRKACA
Genomic Map