Assay Details
| Gene Symbol: TTPAL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002747576.3 | XP_002747622.1 | 3 - 4 | 761 | 88 | |
| XM_008995977.2 | XP_008994225.1 | 3 - 4 | 822 | 88 | ||
More Information
Set Membership: |
|

TTPAL
Genomic Map