Assay Details
Gene Symbol: NSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002746045.4 | XP_002746091.1 | 15 - 16 | 3812 | 106 | |
XM_008993711.2 | XP_008991959.1 | 13 - 14 | 3605 | 106 | ||
XM_017970711.1 | XP_017826200.1 | 15 - 16 | 3482 | 106 | ||
XM_017970712.1 | XP_017826201.1 | 16 - 17 | 3919 | 106 | ||
XM_017970713.1 | XP_017826202.1 | 14 - 15 | 2339 | 106 |

More Information
Set Membership: |
![]() ![]() |
