Assay Details
Gene Symbol: NSD2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002746045.4 | XP_002746091.1 | 16 - 17 | 3912 | 85 | |
XM_008993711.2 | XP_008991959.1 | 14 - 15 | 3705 | 85 | ||
XM_017970711.1 | XP_017826200.1 | 16 - 17 | 3582 | 85 | ||
XM_017970712.1 | XP_017826201.1 | 17 - 18 | 4019 | 85 | ||
XM_017970713.1 | XP_017826202.1 | 15 - 16 | 2439 | 85 |
