Assay Details
| Gene Symbol: NSD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002746045.4 | XP_002746091.1 | 19 - 20 | 4447 | 87 | |
| XM_008993711.2 | XP_008991959.1 | 17 - 18 | 4240 | 87 | ||
| XM_017970711.1 | XP_017826200.1 | 19 - 20 | 4117 | 87 | ||
| XM_017970712.1 | XP_017826201.1 | 20 - 21 | 4554 | 87 | ||
| XM_017970713.1 | XP_017826202.1 | 18 - 19 | 2974 | 87 | ||
More Information
Set Membership: |
|

NSD2
Genomic Map