Assay Details
| Gene Symbol: ALG6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001381.2 | XP_008999629.1 | 9 - 10 | 1118 | 88 | |
| XM_009001382.2 | XP_008999630.1 | 9 - 10 | 1217 | 88 | ||
| XM_009001383.2 | XP_008999631.1 | 9 - 10 | 989 | 88 | ||
| XM_009001384.2 | XP_008999632.1 | 9 - 10 | 1097 | 88 | ||
| XM_017974807.1 | XP_017830296.1 | 10 - 11 | 1157 | 88 | ||
More Information
Set Membership: |
|

ALG6
Genomic Map