Assay Details
| Gene Symbol: KCTD15 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008987703.2 | XP_008985951.1 | 6 - 7 | 965 | 60 | |
| XM_017968040.1 | XP_017823529.1 | 6 - 7 | 965 | 60 | ||
| XM_017968041.1 | XP_017823530.1 | 6 - 7 | 953 | 60 | ||
More Information
Set Membership: |
|

KCTD15
Genomic Map