Assay Details
| Gene Symbol: AQP4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002757138.4 | XP_002757184.1 | 4 - 5 | 838 | 77 | |
| XM_008979641.2 | XP_008977889.1 | 5 - 6 | 862 | 77 | ||
More Information
Set Membership: |
|

AQP4
Genomic Map