Assay Details
| Gene Symbol: CERS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761683.3 | XP_002761729.1 | 7 - 8 | 1062 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CERS4| Gene Symbol: CERS4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002761683.3 | XP_002761729.1 | 7 - 8 | 1062 | 64 | |
Set Membership: |
|