Assay Details
| Gene Symbol: EMG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002752274.4 | XP_002752320.1 | 3 - 4 | 554 | 69 | |
| XM_009003503.2 | XP_009001751.1 | 3 - 4 | 330 | 69 | ||
| XM_017975912.1 | XP_017831401.1 | 3 - 4 | 554 | 69 | ||
| XM_017975913.1 | XP_017831402.1 | 3 - 4 | 371 | 69 | ||
More Information
Set Membership: |
|

EMG1
Genomic Map