Assay Details
| Gene Symbol: CDH13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_008986283.2 | XP_008984531.2 | 9 - 10 | 1574 | 89 | |
| XM_017967057.1 | XP_017822546.1 | 9 - 10 | 1574 | 89 | ||
More Information
Set Membership: |
|

CDH13
Genomic Map