Assay Details
| Gene Symbol: PRPF18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002750048.4 | XP_002750094.1 | 1 - 2 | 336 | 67 | |
| XM_008999958.2 | XP_008998206.1 | 1 - 2 | 216 | 67 | ||
| XM_008999959.2 | XP_008998207.1 | 1 - 2 | 218 | 67 | ||
| XM_008999960.2 | XP_008998208.1 | 1 - 2 | 336 | 67 | ||
More Information
Set Membership: |
|

PRPF18
Genomic Map