Assay Details
| Gene Symbol: PCYT1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002759773.3 | XP_002759819.1 | 7 - 8 | 1247 | 68 | |
| XM_008984116.2 | XP_008982364.1 | 7 - 8 | 888 | 68 | ||
More Information
Set Membership: |
|

PCYT1A
Genomic Map