Assay Details
| Gene Symbol: MYL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749740.4 | XP_002749786.2 | 5 - 6 | 510 | 82 | |
| XM_008999416.1 | XP_008997664.1 | 5 - 6 | 695 | 82 | ||
More Information
Set Membership: |
|

MYL1
Genomic Map