Assay Details
| Gene Symbol: PLA2G12B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002756249.3 | XP_002756295.2 | 4 | 856 | 80 | |
| XM_009009745.1 | XP_009007993.1 | 4 | 853 | 80 | ||
More Information
Set Membership: |
|

PLA2G12B
Genomic Map