Assay Details
| Gene Symbol: CLTA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002743076.3 | XP_002743122.1 | 2 - 3 | 444 | 70 | |
| XM_002743077.3 | XP_002743123.1 | 2 - 3 | 444 | 70 | ||
| XM_002743078.3 | XP_002743124.1 | 2 - 3 | 444 | 70 | ||
| XM_002743079.3 | XP_002743125.1 | 2 - 3 | 444 | 70 | ||
| XM_017974704.1 | XP_017830193.1 | 2 - 3 | 444 | 70 | ||
More Information
Set Membership: |
|

CLTA
Genomic Map