Assay Details
| Gene Symbol: LSMEM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002758375.3 | XP_002758421.1 | 4 - 5 | 777 | 70 | |
| XM_008981678.2 | XP_008979926.1 | 4 - 5 | 774 | 70 | ||
More Information
Set Membership: |
|

LSMEM2
Genomic Map