Assay Details
Gene Symbol: CAPNS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002762038.4 | XP_002762084.1 | 9 - 10 | 984 | 65 | |
XM_008987793.2 | XP_008986041.1 | 9 - 10 | 987 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
