Assay Details
| Gene Symbol: CYP2A13 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_017968083.1 | XP_017823572.1 | 7 - 8 | 1644 | 70 | |
| GenBank mRNA | KJ922555.1 | - | 7 - 8 | 1363 | 70 | |
More Information
Set Membership: |
|

CYP2A13
Genomic Map