Assay Details
| Gene Symbol: ELF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002748997.4 | XP_002749043.1 | 5 - 6 | 2200 | 62 | |
| XM_002748998.4 | XP_002749044.1 | 5 - 6 | 2129 | 62 | ||
| XM_008998179.2 | XP_008996427.1 | 5 - 6 | 1086 | 62 | ||
| XM_008998180.2 | XP_008996428.1 | 5 - 6 | 799 | 62 | ||
More Information
Set Membership: |
|

ELF1
Genomic Map