Assay Details
| Gene Symbol: ALOX12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002806810.3 | XP_002806856.1 | 8 - 9 | 2455 | 60 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

ALOX12| Gene Symbol: ALOX12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002806810.3 | XP_002806856.1 | 8 - 9 | 2455 | 60 | |
Set Membership: |
|