Assay Details
| Gene Symbol: SH3GLB2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009003745.2 | XP_009001993.1 | 10 - 11 | 977 | 64 | |
| XM_009003751.2 | XP_009001999.1 | 9 - 10 | 965 | 64 | ||
| XM_009003766.2 | XP_009002014.1 | 8 - 9 | 902 | 64 | ||
More Information
Set Membership: |
|

SH3GLB2
Genomic Map