Assay Details
| Gene Symbol: PLCL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002749603.4 | XP_002749649.3 | 5 - 6 | 3495 | 76 | |
| XM_017973600.1 | XP_017829089.1 | 4 - 5 | 3291 | 76 | ||
More Information
Set Membership: |
|

PLCL1
Genomic Map