Assay Details
| Gene Symbol: IMMP2L | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751760.4 | XP_002751806.1 | 1 - 2 | 281 | 77 | |
| XM_009002709.2 | XP_009000957.1 | 3 - 4 | 439 | 77 | ||
| XM_009002710.2 | XP_009000958.1 | 281 | 77 | |||
| XM_017975464.1 | XP_017830953.1 | 3 - 4 | 281 | 77 | ||
| XM_017975465.1 | XP_017830954.1 | 1 - 2 | 241 | 77 | ||
| XM_017975466.1 | XP_017830955.1 | 2 - 3 | 492 | 77 | ||
| XM_017975467.1 | XP_017830956.1 | 1 - 2 | 332 | 77 | ||
More Information
Set Membership: |
|

IMMP2L
Genomic Map