Assay Details
Gene Symbol: RIC8B | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_002752943.3 | XP_002752989.1 | 6 - 7 | 1358 | 71 | |
XM_009004537.2 | XP_009002785.1 | 6 - 7 | 1358 | 71 | ||
XM_009004538.2 | XP_009002786.1 | 6 - 7 | 1358 | 71 | ||
XM_017976343.1 | XP_017831832.1 | 5 - 6 | 1310 | 71 | ||
XM_017976344.1 | XP_017831833.1 | 6 - 7 | 1720 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |
