Assay Details
| Gene Symbol: MRPS18C | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002745644.4 | XP_002745690.2 | 2 - 3 | 296 | 97 | |
| XM_008993055.1 | XP_008991303.1 | 2 - 3 | 296 | 97 | ||
More Information
Set Membership: |
|

MRPS18C
Genomic Map