Assay Details
| Gene Symbol: DNAJB4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_009001532.2 | XP_008999780.1 | 2 - 3 | 492 | 120 | |
| XM_009001533.2 | XP_008999781.1 | 2 - 3 | 524 | 120 | ||
More Information
Set Membership: |
|

DNAJB4
Genomic Map