Assay Details
| Gene Symbol: SYT12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002755573.3 | XP_002755619.1 | 5 - 6 | 1691 | 75 | |
| XM_017978306.1 | XP_017833795.1 | 4 - 5 | 1171 | 75 | ||
| XM_017978307.1 | XP_017833796.1 | 5 - 6 | 1681 | 75 | ||
More Information
Set Membership: |
|

SYT12
Genomic Map