Assay Details
| Gene Symbol: UBE2D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751370.3 | XP_002751416.1 | 3 - 4 | 211 | 93 | |
| XM_009002105.2 | XP_009000353.1 | 3 - 4 | 348 | 93 | ||
| XM_009002108.2 | XP_009000356.1 | 4 - 5 | 262 | 93 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_009002108.2 transcripts.
Set Membership: |
|

UBE2D4
Genomic Map