Assay Details
| Gene Symbol: UBE2D4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_002751370.3 | XP_002751416.1 | 5 - 6 | 398 | 60 | |
| XM_009002103.2 | XP_009000351.1 | 6 - 7 | 447 | 60 | ||
| XM_009002104.2 | XP_009000352.1 | 5 - 6 | 557 | 60 | ||
| XM_009002105.2 | XP_009000353.1 | 5 - 6 | 535 | 60 | ||
| XM_009002108.2 | XP_009000356.1 | 6 - 7 | 449 | 60 | ||
| XM_017975151.1 | XP_017830640.1 | 6 - 7 | 443 | 60 | ||
| XM_017975152.1 | XP_017830641.1 | 5 - 6 | 390 | 60 | ||
| XM_017975153.1 | XP_017830642.1 | 6 - 7 | 444 | 60 | ||
More Information
Set Membership: |
|

UBE2D4
Genomic Map